missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)imidazole, 98%

Catalog No. AAB2562106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2562106 5 g
AAB2562103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2562106 Supplier Thermo Scientific Chemicals Supplier No. B2562106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10040-95-6 | C10H10N2O | 174.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10040-95-6
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00060492
InChI Key XNLOIFUGGCCEQX-UHFFFAOYSA-N
PubChem CID 82328
IUPAC Name 1-(4-methoxyphenyl)imidazole
SMILES COC1=CC=C(C=C1)N2C=CN=C2

Specifications

Melting Point 64°C to 67°C
Boiling Point 120°C (0.1 mmHg)
Odor Odorless
Quantity 5 g
Synonym 1-4-methoxyphenyl-1h-imidazole, 1-4-methoxyphenyl imidazole, n-4-methoxyphenyl imidazole, 1h-imidazole, 1-4-methoxyphenyl, 1-4-methoxy-phenyl-1h-imidazole, acmc-20ahwg, 4-imidazol-1-yl anisole, 4-imidazol-1-yl-anisole, 1-imidazolyl-4-methoxybenzene, 1h-imidazole,1-4-methoxyphenyl
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material 1-(4-Methoxyphenyl)imidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only