missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)ethanol, 95%

Catalog No. p-7045810
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1206214 25 g
AAL1206222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1206214 Supplier Thermo Scientific Chemicals Supplier No. L1206214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3319-15-1 | C9H12O2 | 152.19 g/mol

1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

Solubility
Not miscible in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3319-15-1
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016857,MFCD09863666,MFCD09863667
InChI Key IUUULXXWNYKJSL-UHFFFAOYNA-N
Synonym 1-4-methoxyphenyl ethanol, 1-4-methoxyphenyl ethan-1-ol, 4-methoxy-alpha-methylbenzyl alcohol, 4-methoxyphenyl methyl carbinol, 1-p-methoxyphenyl ethanol, 4-methoxy-.alpha.-methylbenzyl alcohol, 1-4-methoxyphenyl-ethanol, acmc-20apae, 4-methoxyphenyl ethanol, 1 p-methoxyphenyl ethanol
PubChem CID 101148
ChEBI CHEBI:86566
IUPAC Name 1-(4-methoxyphenyl)ethanol
SMILES COC1=CC=C(C=C1)C(C)O

Specifications

Density 1.079
Boiling Point 94°C to 96°C (1 mmHg)
Flash Point 74°C (165°F)
Refractive Index 1.533
Quantity 25 g
Beilstein 2043521
Solubility Information Not miscible in water.
Formula Weight 152.19
Percent Purity 95%
Chemical Name or Material 1-(4-Methoxyphenyl)ethanol
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 222-019-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only