missing translation for 'onlineSavingsMsg'
Learn More

1,4-Hexadiene, cis + trans, 99%

Catalog No. AAB2006006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2006006 5 g
AAB2006014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2006006 Supplier Thermo Scientific Chemicals Supplier No. B2006006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 592-45-0 | C6H10 | 82.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 592-45-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009298
InChI Key PRBHEGAFLDMLAL-GQCTYLIASA-N
Synonym 1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene
PubChem CID 5365552
IUPAC Name (4E)-hexa-1,4-diene
SMILES CC=CCC=C

Specifications

Density 0.71
Boiling Point 64°C to 66°C
Flash Point −26°C (−15°F)
Odor Characteristic
Refractive Index 1.411
Quantity 5 g
UN Number UN2458
Beilstein 1731889
Formula Weight 82.15
Percent Purity 99%
Chemical Name or Material 1,4-Hexadiene, cis + trans
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXADIENES
EINECSNumber 209-756-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only