Learn More
Foxx Life Sciences VersaBarb Hose Barb Spigot Fitting, 1/4 Inch, for use with Foxx EZgrip Carboys and Nalgene-Style Containers
Supplier: Foxx Life Sciences 2254002RLS
225-4002-RLS: VersaBarb, Spigot Fitting, 1 1/8" Thread with 1/4" HB. Foxx Life Sciences VersaBarb provides flexibility in drainage, pump connection, filling bell, and automated equipment. VersaBarb is available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes. In addition, a 3/4" sanitary fitting VersaBarb is also available to enable the EZgrip carboy to function as a supply reservoir for a larger system. Offers a convenient solution for bottom-connections and can fit a variety of tubing diameters. Made from Class VI autoclavable polypropylene. Available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes or 3/4" sanitary fitting.