missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenyl-1,4-butanedione 98.0+%, TCI America™

Catalog No. D31791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31791G Supplier TCI America Supplier No. D31791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 495-71-6
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
InChI Key OSWWFLDIIGGSJV-UHFFFAOYSA-N
Synonym 1,2-dibenzoylethane, 1,4-diphenyl-1,4-butanedione, 1,4-butanedione, 1,4-diphenyl, biphenacyl, diphenacyl, 2,2-biacetophenone, acmc-1bd9f, ethane, 1,2-dibenzoyl, succinophenone diphenacyl, cambridge id 5102527
PubChem CID 136322
IUPAC Name 1,4-diphenylbutane-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)CCC(=O)C2=CC=CC=C2

Specifications

Melting Point 148°C
Color White
Boiling Point 260°C
Quantity 1 g
Formula Weight 238.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diphenyl-1,4-butanedione