missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane (stabilized with BHT) 99.0+%, TCI America™
$37.29 - $54.06
Chemical Identifiers
| CAS | 123-91-1 |
|---|---|
| Molecular Formula | C4H8O2 |
| Molecular Weight (g/mol) | 88.106 |
| MDL Number | MFCD00006571 |
| InChI Key | RYHBNJHYFVUHQT-UHFFFAOYSA-N |
| Synonym | dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin |
| PubChem CID | 31275 |
| ChEBI | CHEBI:47032 |
| IUPAC Name | 1,4-dioxane |
| SMILES | C1COCCO1 |
Chemical Identifiers
| 123-91-1 | |
| 88.106 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 31275 | |
| 1,4-dioxane |
| C4H8O2 | |
| MFCD00006571 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| CHEBI:47032 | |
| C1COCCO1 |
Specifications
| 123-91-1 | |
| 101°C | |
| Clear Liquid at 20°C | |
| MFCD00006571 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| C1COCCO1 | |
| 88.106 | |
| CHEBI:47032 | |
| ≥99.0% (GC) |
| Colorless | |
| 25 g | |
| C4H8O2 | |
| 1165 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 1,4-dioxane | |
| 31275 | |
| 88.11 | |
| 1,4-Dioxane (stabilized with BHT) |
Safety and Handling
EINECSNumber : (5)-0839
RTECSNumber : JG8225000
TSCA : Yes