missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane (stabilized with BHT) 99.0+%, TCI America™

Catalog No. D0860500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0860500G 500 g
D086025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0860500G Supplier TCI America Supplier No. D0860500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Color Colorless
Quantity 500 g
Physical Form Clear Liquid at 20°C
UN Number 1165
Boiling Point 101°C
Formula Weight 88.11
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1,4-Dioxane (stabilized with BHT)
EINECSNumber (5)-0839
RTECSNumber JG8225000
TSCA Yes