missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.5% (GC), Solstice

Catalog No. 60046209
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
210 kg
Packaging:
Glass Bottle
Metal Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046209 4 x 2.5 L Glass Bottle
60046206 1 L Glass Bottle
60046207 2.5 L Glass Bottle
60046208 210 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60046209 Supplier Solstice Supplier No. 331474X2.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

pH 6 to 8 (at 500g/L 20°C)
Melting Point 10°C to 12°C
Color Colorless
Density 1.034g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.22
Flash Point 12°C (53.6°F)
Ignition Point 356°F
Odor Aromatic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 40mmHg (25°C), 27mmHg (20°C)
UN Number UN1165
Beilstein 102551
Boiling Point 100°C to 102°C
Formula Weight 88.11g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 1,4-Dioxane, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P261-P281-P305 + P351 + P338
Precautionary Statement H225-H319-H335-H351
DOTInformation class 3 - PG 2 - Dioxane
EINECSNumber 204-661-8
RTECSNumber JG8225000