missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.5% (GC), Solstice
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| pH | 6 to 8 (at 500g/L 20°C) |
| Melting Point | 10°C to 12°C |
| Color | Colorless |
| Density | 1.034g/mL at 25°C |
| Quantity | 210 kg |
| Explosion Limit | 0.22 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 356°F |
| Odor | Aromatic |
| Packaging | Metal Drum |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | P210-P261-P281-P305 + P351 + P338 |
| Precautionary Statement | H225-H319-H335-H351 |
| DOTInformation | class 3 - PG 2 - Dioxane |
| EINECSNumber | 204-661-8 |
| RTECSNumber | JG8225000 |