missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, Honeywell™

Catalog No. 6000075
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000075 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000075 Supplier Honeywell Chemicals Supplier No. 349441L
Only null left

Puriss. p.a., dried,≥99.5% (GC), ≤0.005% water

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Density 1.034g/mL at 25°C
Quantity 1 L
Explosion Limit 0.22
Flash Point 12°C (53.6°F)
Ignition Point 356°F
Vapor Pressure 27 mmHg (20°C), 40 mmHg (25°C)
UN Number UN1165
Beilstein 102551
Formula Weight 88.11g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material 1,4-Dioxane
Show More Show Less
Hazard Statement P210-P261-P281-P305 + P351 + P338
RTECSNumber JG8225000