missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane (Certified ACS), Fisher Chemical™

Catalog No. D1114
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D1114 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1114 Supplier Thermo Fisher Scientific Supplier No. BPD1114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

pH 6 to 8
Melting Point 12°C
Color Colorless
Density 1.034g/cm³
Quantity 4 L
Flash Point 12°C (53.6°F)
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.004% max.
Assay Percent Range ≥99 %
Titratable Acid 0.0016mEq/g max.
Vapor Pressure 41mbar at 20°C
Viscosity 1.32 mPa/s at 20°C
Water 0.02% max.
Boiling Point 101°C
Formula Weight 88.11
Percent Purity ≥99%
Grade ACS
Chemical Name or Material 1, 4-Dioxane
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Category SIGNAL WORD: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. Inhalation may cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Hygroscopic.
NFPA
Health: 2
Flammability: 3
Instability: 1
DOTInformation DOT Class 3, : Flammable Liquid