missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99+%, stab. with ca 5-10ppm BHT

Catalog No. p-7021732
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1418836 500 g
AAA141880E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1418836 Supplier Thermo Scientific Chemicals Supplier No. A1418836
Only null left

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol

1,4-Dioxane is used as a stabilizer for 1,1,1-trichloroethane and chlorinated solvents. It acts as a solvent for cellulose acetate, ethyl cellulose, benzyl cellulose, resins, oils, waxes, dyes, other organic and inorganic compounds. It acts as a substitute for tetrahydrofuran due to its higher boiling point and lower toxicity. Further, it is used as an internal standard for proton NMR spectroscopy in deuterium oxide. It plays an important role in liquid scintillation counting in biological procedures, and as a wetting and dispersing agent in textile processing. It is involved in the preparation of dimethylmagnesium by reacting with methylmagnesium bromide, and in the preparation of histological sections for microscopic examination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dioxane is used as a stabilizer for 1,1,1-trichloroethane and chlorinated solvents. It acts as a solvent for cellulose acetate, ethyl cellulose, benzyl cellulose, resins, oils, waxes, dyes, other organic and inorganic compounds. It acts as a substitute for tetrahydrofuran due to its higher boiling point and lower toxicity. Further, it is used as an internal standard for proton NMR spectroscopy in deuterium oxide. It plays an important role in liquid scintillation counting in biological procedures, and as a wetting and dispersing agent in textile processing. It is involved in the preparation of dimethylmagnesium by reacting with methylmagnesium bromide, and in the preparation of histological sections for microscopic examination.

Solubility
Miscible with water, organic solvents, aromatic hydrocarbons and oils.

Notes
Hygroscopic. Incompatible with oxygen, oxidizing agents, halogens, reducing agents, perchlorates and trimethylaluminum.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 11.8°C
Density 1.034
Boiling Point 100°C to 102°C
Quantity 500 g
Odor Ether-like
Refractive Index 1.422
UN Number UN1165
Beilstein 102551
Merck Index 14,3300
Solubility Information Miscible with water,organic solvents,aromatic hydrocarbons and oils.
Formula Weight 88.11
Percent Purity ≥99%
Chemical Name or Material 1,4-Dioxane, stabilized with ≈5 to 10ppm BHT
Show More Show Less
Hazard Category H225-H319-H335-H336-H350-H373
Hazard Statement GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapour.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIOXANE
EINECSNumber 204-661-8
RTECSNumber JG8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only