Learn More
1,4-Dioxane, 99+%, for spectroscopy
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Supplier: Thermo Scientific Chemicals 167800025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
| 1, 4-Dioxane | |
| 99+% | |
| 0.01 max. at 295nm, 0.03 max. at 270nm, 0.05 max. at 260nm, 0.05 max. at 275nm, 0.2 max. at 250nm, 0.5 max. at 225nm, 0.5 max. at 230nm, 1 max. at 215nm, 1 max. at 220nm | |
| 99.0 | |
| 1.0340g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.4205 to 1.4220 | |
| 99+% | |
| C4H8O2 | |
| 1.32 mPa.s (20°C) | |
| 19, 3 | |
| 15, 3328 | |
| Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents | |
| C1COCCO1 | |
| 88.106 | |
| CHEBI:47032 | |
| ≥99% |
| 12.0°C | |
| 123-91-1 | |
| White | |
| 100.0 | |
| 101.0°C | |
| 12°C | |
| Crystalline Powder, Crystals, and/or Chunks | |
| 0.0005 % max. | |
| 1.034 | |
| MFCD00006571 | |
| 0.03% Max. | |
| 01,333 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 1,4-dioxane | |
| 31275 | |
| 88.11 | |
| Spectroscopy |
Chemical Identifiers
| 123-91-1 | |
| 88.106 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 31275 | |
| 1,4-dioxane |
| C4H8O2 | |
| MFCD00006571 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| CHEBI:47032 | |
| C1COCCO1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 204-661-8
RTECSNumber : JG8225000
TSCA : TSCA
RUO – Research Use Only