missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99%, for biochemistry, stabilized, AcroSeal™

Catalog No. AC447060025
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447060025 2.5 L AcroSeal™ Glass Bottle
AC447060010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447060025 Supplier Thermo Scientific Chemicals Supplier No. 447060025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 12.0°C
Absorbance 0.05 max. at 295nm, 0.1 max. at 275nm, 0.5 max. at 250nm
Density 1.0340g/mL
Quantity 2.5 L
Flash Point 12°C
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4205 to 1.4220
Specific Gravity 1.034
Assay Percent Range 99%
Viscosity 1.32 mPa.s (20°C)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Boiling Point 101.0°C
Formula Weight 88.11
Percent Purity 99%
Grade Biochemical
Chemical Name or Material 1, 4-Dioxane
Show More Show Less
Hazard Category Danger

RUO – Research Use Only