Learn More
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur.
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Supplier: Thermo Scientific Chemicals 408820100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1, 4-Dioxane | |
| 123-91-1 | |
| 100.0 | |
| 101.0°C | |
| 12°C | |
| Liquid | |
| 0.005% max. | |
| 1.034 | |
| C4H8O2 | |
| 1.32 mPa.s (20°C) | |
| 19, 3 | |
| 15, 3328 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 1,4-dioxane | |
| 31275 | |
| 88.11 | |
| ACS Reagent |
| 12.0°C | |
| 99.0 | |
| 1.0340g/mL | |
| 10 L | |
| Plastic Drum | |
| 1.4205 to 1.4220 | |
| 99+% | |
| 0.0016 meq/g max. | |
| MFCD00006571 | |
| 0.05% max. (Coulometric) | |
| 01,333 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| C1COCCO1 | |
| 88.106 | |
| CHEBI:47032 | |
| ≥99% |
Chemical Identifiers
| 123-91-1 | |
| 88.106 | |
| RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
| 31275 | |
| 1,4-dioxane |
| C4H8O2 | |
| MFCD00006571 | |
| dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
| CHEBI:47032 | |
| C1COCCO1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. Inhalation may cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Hygroscopic.
NFPA
Health: 2
Flammability: 3
Instability: 1
SIGNAL WORD: DANGER!
EINECSNumber : 204-661-8
RTECSNumber : JG8225000
TSCA : TSCA
RUO – Research Use Only