missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxan-2-one 98.0+%, TCI America™

Catalog No. D46445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46445G 5 g
D464425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46445G Supplier TCI America Supplier No. D46445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3041-16-5
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00215855
InChI Key VPVXHAANQNHFSF-UHFFFAOYSA-N
Synonym p-Dioxanone
PubChem CID 18233
IUPAC Name 1,4-dioxan-2-one
SMILES C1COC(=O)CO1

Specifications

Melting Point 28°C
Color White
Quantity 5 g
Physical Form Crystal-Lump at 20°C
Boiling Point 215°C
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,4-Dioxan-2-one
RTECSNumber JH2290000
TSCA No
Recommended Storage Refrigerator