missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dimethylnaphthalene 96.0+%, TCI America™
$257.24 - $904.06
Chemical Identifiers
| CAS | 571-58-4 |
|---|---|
| Molecular Formula | C12H12 |
| Molecular Weight (g/mol) | 156.228 |
| MDL Number | MFCD00004037 |
| InChI Key | APQSQLNWAIULLK-UHFFFAOYSA-N |
| Synonym | naphthalene, 1,4-dimethyl, 1,4-dimethylnapthalene, unii-bqh3s1i0so, 1,4-dmn, 1,4-dimethyl-naphthalene, bqh3s1i0so, epa pesticide chemical code 055802, naphthalene,4-dimethyl, naphthalene,1,4-dimethyl, acmc-1b0g8 |
| PubChem CID | 11304 |
| ChEBI | CHEBI:48609 |
| IUPAC Name | 1,4-dimethylnaphthalene |
| SMILES | CC1=CC=C(C2=CC=CC=C12)C |
Chemical Identifiers
| 571-58-4 | |
| 156.228 | |
| APQSQLNWAIULLK-UHFFFAOYSA-N | |
| 11304 | |
| 1,4-dimethylnaphthalene |
| C12H12 | |
| MFCD00004037 | |
| naphthalene, 1,4-dimethyl, 1,4-dimethylnapthalene, unii-bqh3s1i0so, 1,4-dmn, 1,4-dimethyl-naphthalene, bqh3s1i0so, epa pesticide chemical code 055802, naphthalene,4-dimethyl, naphthalene,1,4-dimethyl, acmc-1b0g8 | |
| CHEBI:48609 | |
| CC1=CC=C(C2=CC=CC=C12)C |
Specifications
| 571-58-4 | |
| 268°C | |
| MFCD00004037 | |
| naphthalene, 1,4-dimethyl, 1,4-dimethylnapthalene, unii-bqh3s1i0so, 1,4-dmn, 1,4-dimethyl-naphthalene, bqh3s1i0so, epa pesticide chemical code 055802, naphthalene,4-dimethyl, naphthalene,1,4-dimethyl, acmc-1b0g8 | |
| CC1=CC=C(C2=CC=CC=C12)C | |
| 156.228 | |
| CHEBI:48609 | |
| ≥96.0% (GC) | |
| 1,4-Dimethylnaphthalene |
| Yellow | |
| C12H12 | |
| 5 g | |
| APQSQLNWAIULLK-UHFFFAOYSA-N | |
| 1,4-dimethylnaphthalene | |
| 11304 | |
| 156.23 | |
| Liquid |
Safety and Handling
EINECSNumber : (4)-0080
RTECSNumber : QJ4440000
TSCA : No