missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dimethylnaphthalene 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 268°C |
| Quantity | 5 g |
| Formula Weight | 156.23 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,4-Dimethylnaphthalene |
Safety and Handling
| EINECSNumber | (4)-0080 |
| RTECSNumber | QJ4440000 |
| TSCA | No |