missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethylnaphthalene 96.0+%, TCI America™

Catalog No. D100625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D100625G 25 g
D10065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D100625G Supplier TCI America Supplier No. D100625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 571-58-4
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004037
InChI Key APQSQLNWAIULLK-UHFFFAOYSA-N
Synonym naphthalene, 1,4-dimethyl, 1,4-dimethylnapthalene, unii-bqh3s1i0so, 1,4-dmn, 1,4-dimethyl-naphthalene, bqh3s1i0so, epa pesticide chemical code 055802, naphthalene,4-dimethyl, naphthalene,1,4-dimethyl, acmc-1b0g8
PubChem CID 11304
ChEBI CHEBI:48609
IUPAC Name 1,4-dimethylnaphthalene
SMILES CC1=CC=C(C2=CC=CC=C12)C

Specifications

Color Yellow
Boiling Point 268°C
Quantity 25 g
Formula Weight 156.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dimethylnaphthalene
EINECSNumber (4)-0080
RTECSNumber QJ4440000
TSCA No