missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethylcyclohexane (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7126383
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D070025ML 25 mL
D0700500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D070025ML Supplier TCI America Supplier No. D070025ML
Only null left

Chemical Identifiers

CAS 589-90-2
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00001508,MFCD00064174,MFCD00064955
InChI Key QRMPKOFEUHIBNM-UHFFFAOYSA-N
Synonym Hexahydro-p-xylene
PubChem CID 11523
IUPAC Name 1,4-dimethylcyclohexane
SMILES CC1CCC(C)CC1

Specifications

Color Colorless
Quantity 25 mL
UN Number 2263
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dimethylcyclohexane (cis- and trans- mixture)
EINECSNumber (3)-2227
TSCA No