missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethyl-1,2,4-triazolium Iodide 98.0+%, TCI America™

$147.25 - $438.05

Chemical Identifiers

CAS 120317-69-3
Molecular Formula C4H8IN3
Molecular Weight (g/mol) 225.033
InChI Key IMFBMMNQDMTHGG-UHFFFAOYSA-M
PubChem CID 10987916
IUPAC Name 1,4-dimethyl-1,2,4-triazol-4-ium;iodide
SMILES CN1C=[N+](C=N1)C.[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D39621G
View Documents
TCI America
D39621G
1 g
Each for $147.25
Only null left
 
D39625G
View Documents
TCI America
D39625G
5 g
Each for $438.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

120317-69-3
225.033
10987916
CN1C=[N+](C=N1)C.[I-]
C4H8IN3
IMFBMMNQDMTHGG-UHFFFAOYSA-M
1,4-dimethyl-1,2,4-triazol-4-ium;iodide

Specifications

120317-69-3
Yellow
1 g
CN1C=[N+](C=N1)C.[I-]
225.033
225.03
Crystalline Powder
118°C
C4H8IN3
IMFBMMNQDMTHGG-UHFFFAOYSA-M
1,4-dimethyl-1,2,4-triazol-4-ium;iodide
10987916
≥98.0% (HPLC,T)
1,4-Dimethyl-1,2,4-triazolium Iodide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'