missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 97%, Thermo Scientific™

Catalog No. p-3881225
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC37116DA 1g
CC37116DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC37116DA Supplier Thermo Scientific Chemicals Supplier No. CC37116DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 857283-87-5
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD09702356
InChI Key NBANRNWRIHAGBJ-UHFFFAOYSA-N
Synonym 1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
PubChem CID 24229481
IUPAC Name 1,4-dimethyl-2,3-dihydroquinoxaline-6-carbonitrile
SMILES CN1CCN(C2=C1C=CC(=C2)C#N)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material 1,4-Dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.