missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxynaphthalene, 96%

CAS: 10075-62-4 | C12H12O2 | 188.226 g/mol

Supplier:  Thermo Scientific Chemicals A1409514

 View more versions of this product

Catalog No. AAA1409514


Only null left
Explore available promotions

Description

Description

1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1,4-Dimethoxynaphthalene
84°C to 85°C
MFCD00052378
2048082
Soluble in water.
COC1=CC=C(C2=CC=CC=C21)OC
188.226
188.23
10075-62-4
C12H12O2
25 g
naphthalene, 1,4-dimethoxy, 1,4-dimethoxy-naphthalene, maybridge1_003006, naphthalene,4-dimethoxy, acmc-1c6fs, 1,4-dimethoxy naphthalene, naphthalene,1,4-dimethoxy, 1,4-dimethoxynaphthalene, 1,4-dimethoxynaphthalene, for fluorescence, fluorescent marker hplc
FWWRTYBQQDXLDD-UHFFFAOYSA-N
1,4-dimethoxynaphthalene
82337
96%

Chemical Identifiers

10075-62-4
188.226
FWWRTYBQQDXLDD-UHFFFAOYSA-N
1,4-dimethoxynaphthalene
C12H12O2
MFCD00052378
82337
COC1=CC=C(C2=CC=CC=C21)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 233-209-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only