missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxynaphthalene, 96%

Catalog No. AAA1409503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1409503 1 g
AAA1409506 5 g
AAA1409514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1409503 Supplier Thermo Scientific Chemicals Supplier No. A1409503

CAS: 10075-62-4 | C12H12O2 | 188.226 g/mol

1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10075-62-4
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00052378
InChI Key FWWRTYBQQDXLDD-UHFFFAOYSA-N
PubChem CID 82337
IUPAC Name 1,4-dimethoxynaphthalene
SMILES COC1=CC=C(C2=CC=CC=C21)OC

Specifications

Melting Point 84°C to 85°C
Quantity 1 g
Beilstein 2048082
Synonym naphthalene, 1,4-dimethoxy, 1,4-dimethoxy-naphthalene, maybridge1_003006, naphthalene,4-dimethoxy, acmc-1c6fs, 1,4-dimethoxy naphthalene, naphthalene,1,4-dimethoxy, 1,4-dimethoxynaphthalene, 1,4-dimethoxynaphthalene, for fluorescence, fluorescent marker hplc
Solubility Information Soluble in water.
Formula Weight 188.23
Percent Purity 96%
Chemical Name or Material 1,4-Dimethoxynaphthalene
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 233-209-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only