Learn More
1,4-Diisocyanatobutane, 97%
CAS: 4538-37-8 | C6H8N2O2 | 140.14 g/mol
Supplier: Thermo Scientific Chemicals 314380050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1, 4-Diisocyanatobutane | |
| 96.0 | |
| Yellow | |
| 102.0°C to 104.0°C (14.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4510 to 1.4530 | |
| MFCD00075559 | |
| 2206 | |
| 1,4-butanediisocyanate, tetramethylene diisocyanate, 1,4-disocyanatobutane, acmc-1afua, 1,4-diisocyanato-butane, butane-1,4-diisocyanate, #, butane,1,4-diisocyanato, 1,4-butanediyl diisocyanate, butane, 1,4-diisocyanato | |
| C(CCN=C=O)CN=C=O | |
| 140.14 | |
| 140.14 | |
| Liquid |
| 4538-37-8 | |
| 100.0 | |
| 1.1000g/mL | |
| 106°C | |
| 96% min. (GC) | |
| C6H8N2O2 | |
| OCN(CH2)4NCO | |
| 5 g | |
| 1.1 | |
| OVBFMUAFNIIQAL-UHFFFAOYSA-N | |
| 1,4-diisocyanatobutane | |
| 350453 | |
| 97% |
Chemical Identifiers
| 4538-37-8 | |
| 140.14 | |
| OVBFMUAFNIIQAL-UHFFFAOYSA-N | |
| 1,4-diisocyanatobutane |
| C6H8N2O2 | |
| MFCD00075559 | |
| 350453 | |
| C(CCN=C=O)CN=C=O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comforta
GHS Signal Word: Danger
RUO – Research Use Only