missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisocyanatobutane, 97%

Catalog No. AC314380010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314380010 1 g Glass bottle
AC314380050 5 g Glass bottle
AC314380250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC314380010 Supplier Thermo Scientific Chemicals Supplier No. 314380010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4538-37-8 | C6H8N2O2 | 140.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4538-37-8
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00075559
InChI Key OVBFMUAFNIIQAL-UHFFFAOYSA-N
PubChem CID 350453
IUPAC Name 1,4-diisocyanatobutane
SMILES C(CCN=C=O)CN=C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 102.0°C to 104.0°C (14.0 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Linear Formula OCN(CH2)4NCO
Quantity 1 g
UN Number 2206
Specific Gravity 1.1
Synonym 1,4-butanediisocyanate, tetramethylene diisocyanate, 1,4-disocyanatobutane, acmc-1afua, 1,4-diisocyanato-butane, butane-1,4-diisocyanate, #, butane,1,4-diisocyanato, 1,4-butanediyl diisocyanate, butane, 1,4-diisocyanato
Formula Weight 140.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 4-Diisocyanatobutane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comforta

RUO – Research Use Only