missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diiodo-2,3,5,6-tetramethylbenzene 95.0+%, TCI America™

Catalog No. p-7127075
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20285G 5 g
D202825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20285G Supplier TCI America Supplier No. D20285G

Chemical Identifiers

CAS 3268-21-1
Molecular Formula C10H12I2
Molecular Weight (g/mol) 386.02
MDL Number MFCD00142554
InChI Key YSKCEXICRGEWDI-UHFFFAOYSA-N
Synonym 3,6-Diiododurene
PubChem CID 5182526
IUPAC Name 1,4-diiodo-2,3,5,6-tetramethylbenzene
SMILES CC1=C(C)C(I)=C(C)C(C)=C1I

Specifications

Melting Point 140°C
Color White
Quantity 5 g
Formula Weight 386.01
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diiodo-2,3,5,6-tetramethylbenzene