missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydroxyanthraquinone, 95%

Catalog No. p-7081161
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1101030 250 g
AAA110100B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1101030 Supplier Thermo Scientific Chemicals Supplier No. A1101030
Only null left

CAS: 81-64-1 | C14H8O4 | 240.214 g/mol

1,4-Dihydroxyanthraquinone is used to color gasoline and some heating oils. It is used as an intermediate for the synthesis of indanthrene- and alizarin-derived dyes. It is also used to form lake pigments with calcium, barium and lead. It may be used in the synthesis of cyclopentanoids by cyclization of α,β-unsaturated aldehyde with 1, 4 1,4-Dihydroxyanthraquinone. It has been studied as a long range emissive ratiometric fluorescent probe for live cell imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dihydroxyanthraquinone is used to color gasoline and some heating oils. It is used as an intermediate for the synthesis of indanthrene- and alizarin-derived dyes. It is also used to form lake pigments with calcium, barium and lead. It may be used in the synthesis of cyclopentanoids by cyclization of α,β-unsaturated aldehyde with 1, 4 1,4-Dihydroxyanthraquinone. It has been studied as a long range emissive ratiometric fluorescent probe for live cell imaging.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 81-64-1
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001209
InChI Key GUEIZVNYDFNHJU-UHFFFAOYSA-N
Synonym quinizarin, 1,4-dihydroxyanthraquinone, quinizarine, chinizarin, solvent orange 86, 1,4-dihydroxyanthrachinon, smoke orange r, 1,4-dihydroxy-9,10-anthraquinone, macrolex orange gg, 9,10-anthracenedione, 1,4-dihydroxy
PubChem CID 6688
ChEBI CHEBI:37487
IUPAC Name 1,4-dihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O

Specifications

Melting Point 195°C to 198°C
Boiling Point ∼450°C
Flash Point 222°C (431°F)
Quantity 250 g
Beilstein 1914036
Merck Index 14,8064
Solubility Information Soluble in water.
Formula Weight 240.21
Percent Purity 95%
Chemical Name or Material 1,4-Dihydroxyanthraquinone
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 201-368-7
RTECSNumber CB6600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only