missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dicyanonaphthalene 98.0+%, TCI America™

Catalog No. D27585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27585G Supplier TCI America Supplier No. D27585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3029-30-9
Molecular Formula C12H6N2
Molecular Weight (g/mol) 178.194
MDL Number MFCD03093626
InChI Key BENSWQOUPJQWMU-UHFFFAOYSA-N
Synonym 1,4-dicyanonaphthalene, 1,4-naphthalenedicarbonitrile, 1,4-dicyanonahthalene, acmc-209heb, ghl.pd_mitscher_leg0.246
PubChem CID 76414
IUPAC Name naphthalene-1,4-dicarbonitrile
SMILES C1=CC=C2C(=CC=C(C2=C1)C#N)C#N

Specifications

Melting Point 208°C
Color Yellow
Quantity 5 g
UN Number 3439
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dicyanonaphthalene