missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dicyanobenzene, 98%

Catalog No. AC113825000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113825000 500 g Glass bottle
AC113820250 25 g Glass bottle
AC113821000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC113825000 Supplier Thermo Scientific Chemicals Supplier No. 113825000

CAS: 623-26-7 | C8H4N2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-26-7
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001810
InChI Key BHXFKXOIODIUJO-UHFFFAOYSA-N
Synonym terephthalonitrile, 1,4-dicyanobenzene, 1,4-benzenedicarbonitrile, p-phthalodinitrile, 4-cyanobenzonitrile, p-dicyanobenzene, p-benzenedinitrile, p-pdn, terephthalodinitrile, 1,4-benzodinitrile
PubChem CID 12172
IUPAC Name benzene-1,4-dicarbonitrile
SMILES C1=CC(=CC=C1C#N)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 222°C to 227°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CN)2
Quantity 500 g
Beilstein 09, 846
Solubility Information Solubility in water: 0.08g/L (23°C). Other solubilities: soluble 15.3g/L in acetone (20°C),soluble in methanol
Formula Weight 128.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 4-Dicyanobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 210-783-2

RUO – Research Use Only