missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorophthalazine 98.0+%, TCI America™

Catalog No. D37495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37495G 5 g
D37491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37495G Supplier TCI America Supplier No. D37495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10-7-4752
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.03
MDL Number MFCD00006909
InChI Key ODCNAEMHGMYADO-UHFFFAOYSA-N
Synonym phthalazine, 1,4-dichloro, ccris 6799, 1,4-dichloro-phthalazine, unii-0ow60266ko, 1,4-dichlorophthalazine, dichlorophthalazine, 1,4-dichloropthalazine, 1,4-dichorophthalazine, pubchem23238, 1,4-dichlorophtalazine
PubChem CID 78490
IUPAC Name 1,4-dichlorophthalazine
SMILES C1=CC=C2C(=C1)C(=NN=C2Cl)Cl

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 5 g
Formula Weight 199.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dichlorophthalazine