missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorophthalazine, 98%, Thermo Scientific Chemicals

Catalog No. AAH3329506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3329506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3329506 Supplier Thermo Scientific Chemicals Supplier No. H3329506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10-7-4752 | C8H4Cl2N2 | 199.03 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10-7-4752
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.03
MDL Number MFCD00006909
InChI Key ODCNAEMHGMYADO-UHFFFAOYSA-N
Synonym phthalazine, 1,4-dichloro, ccris 6799, 1,4-dichloro-phthalazine, unii-0ow60266ko, 1,4-dichlorophthalazine, dichlorophthalazine, 1,4-dichloropthalazine, 1,4-dichorophthalazine, pubchem23238, 1,4-dichlorophtalazine
PubChem CID 78490
IUPAC Name 1,4-dichlorophthalazine
SMILES C1=CC=C2C(=C1)C(=NN=C2Cl)Cl

Specifications

Melting Point 164°C to 166°C
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 199.04
Percent Purity 98%
Chemical Name or Material 1,4-Dichlorophthalazine
EINECSNumber 225-275-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only