missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dichlorobenzene-D4, SPEX CertiPrep™
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,4-Dichlorobenzene-d4 | 3855-82-1 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | 2mL Amber Prescored Glass Ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| CAS Min % | 0.1, 99.9 |
| CAS Max % | 0.1, 99.9 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Methanol-P and T |
| Quantity | 1 mL |
| Chemical Name or Material | 1,4-Dichlorobenzene-d4 |
Safety and Handling
| DOTInformation | Nonhazardous |
| Recommended Storage | Ambient |
Product intended for laboratory use only