missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, 97%

Catalog No. AAL1483506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1483506 5 g
AAL1483503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1483506 Supplier Thermo Scientific Chemicals Supplier No. L1483506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 202823-67-4 | C10H10Cl2N2 | 229.104 g/mol

1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 202823-67-4
Molecular Formula C10H10Cl2N2
Molecular Weight (g/mol) 229.104
MDL Number MFCD01075667
InChI Key GZIFYLGNTOAIJJ-UHFFFAOYSA-N
Synonym 1,4-dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine, acmc-20ap2h, 5,8-ethanophthalazine, 1,4-dichloro-5,6,7,8-tetrahydro, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0<2,7> dodeca-2 7 ,3,5-triene, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0^ 2,7 dodeca-2,4,6-triene, 1s,8s-3,6-dichloro-4,5-diazatricyclo 6.2.2.0 2 ,? dodeca-2,4,6-triene, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0 2 ,? dodeca-2 7 ,3,5-triene, 3,6-dichloro-4,5-diazatricyclo 6.2.2.0 2 ,? dodeca-2,4,6-triene
PubChem CID 6416107
SMILES C1CC2CCC1C3=C2C(=NN=C3Cl)Cl

Specifications

Melting Point 208°C to 211°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 229.11
Percent Purity 97%
Chemical Name or Material 1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only