missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dichloro-2-butyne, 99%
CAS: 821-10-3 | C4H4Cl2 | 122.98 g/mol
$673.06 - $673.06
Chemical Identifiers
| CAS | 821-10-3 |
|---|---|
| Molecular Formula | C4H4Cl2 |
| Molecular Weight (g/mol) | 122.98 |
| MDL Number | MFCD00000981 |
| InChI Key | RCHDLEVSZBOHOS-UHFFFAOYSA-N |
| Synonym | 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference |
| PubChem CID | 13182 |
| IUPAC Name | 1,4-dichlorobut-2-yne |
| SMILES | ClCC#CCCl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC113281000
|
Thermo Scientific Chemicals
113281000 |
100 mL | Glass bottle |
Each for $673.06
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
| 821-10-3 | |
| 122.98 | |
| RCHDLEVSZBOHOS-UHFFFAOYSA-N | |
| 13182 | |
| ClCC#CCCl |
| C4H4Cl2 | |
| MFCD00000981 | |
| 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference | |
| 1,4-dichlorobut-2-yne |
Specifications
| 821-10-3 | |
| 165°C to 168°C | |
| 99% | |
| C4H4Cl2 | |
| ClCH2C∽CCH2Cl | |
| 100 mL | |
| 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference | |
| ClCC#CCCl | |
| 122.98 | |
| 122.98 | |
| 1, 4-Dichloro-2-butyne, 99% |
| 1.2500g/mL | |
| 160°C | |
| Glass bottle | |
| 1.5030 to 1.506 | |
| MFCD00000981 | |
| 1.25 | |
| RCHDLEVSZBOHOS-UHFFFAOYSA-N | |
| 1,4-dichlorobut-2-yne | |
| 13182 | |
| 99% |
Safety and Handling
LIGHT SENSITIVE
GHS Signal Word: Danger
EINECSNumber : 212-474-8
RTECSNumber : ER9600000
TSCA : TSCA
RUO – Research Use Only