missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-butyne, 99%

Catalog No. AC113281000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113281000 100 mL Glass bottle
AC113280250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113281000 Supplier Thermo Scientific Chemicals Supplier No. 113281000

CAS: 821-10-3 | C4H4Cl2 | 122.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 821-10-3
Molecular Formula C4H4Cl2
Molecular Weight (g/mol) 122.98
MDL Number MFCD00000981
InChI Key RCHDLEVSZBOHOS-UHFFFAOYSA-N
Synonym 1,4-dichloro-2-butyne, 2-butyne, 1,4-dichloro, 1,4-dichlorobutyne, 2-butyne,4-dichloro, 1,4 dichloro-2-butyne, 1,4-dichloro-2-butine, 1,4-dichloro-but-2-yne, acmc-209pn5, wln: g2uu2g, 4-01-00-00973 beilstein handbook reference
PubChem CID 13182
IUPAC Name 1,4-dichlorobut-2-yne
SMILES ClCC#CCCl

Specifications

Density 1.2500g/mL
Boiling Point 165°C to 168°C
Flash Point 160°C
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5030 to 1.506
Linear Formula ClCH2C∽CCH2Cl
Quantity 100 mL
Specific Gravity 1.25
Formula Weight 122.98
Percent Purity 99%
Chemical Name or Material 1, 4-Dichloro-2-butyne, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement LIGHT SENSITIVE
EINECSNumber 212-474-8
RTECSNumber ER9600000
TSCA TSCA

RUO – Research Use Only