missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-butanol 97.0+%, TCI America™

Catalog No. D219410G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D219410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D219410G Supplier TCI America Supplier No. D219410G
Only null left

Chemical Identifiers

CAS 2419-74-1
Molecular Formula C4H8Cl2O
Molecular Weight (g/mol) 143.007
MDL Number MFCD00191343
InChI Key CKNNDWZSFAPUJS-UHFFFAOYSA-N
PubChem CID 17021
IUPAC Name 1,4-dichlorobutan-2-ol
SMILES C(CCl)C(CCl)O

Specifications

Color Yellow
Quantity 10 g
Formula Weight 143.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dichloro-2-butanol