missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibenzyl L-Aspartate p-Toluenesulfonate 97.0+%, TCI America™

Catalog No. C27115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27115G 5 g
C271125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27115G Supplier TCI America Supplier No. C27115G

Chemical Identifiers

CAS 2886-33-1
Molecular Formula C25H27NO7S
Molecular Weight (g/mol) 485.55
MDL Number MFCD00065188
InChI Key HLMUYZYLPUHSNV-UHFFFAOYNA-N
Synonym h-asp obzl-obzl.tosoh, h-asp obzl-obzl tosoh, l-aspartic acid dibenzyl ester tosylate, s-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, l-aspartic acid dibenzyl ester p-toluenesulfonate salt, l-aspartic acid dibenzyl ester 4-toluenesulfonate, h-asp obzl-obzl inverted exclamation mark currencytosoh, h-asp obzl-obzl.tos, l-aspartic acid dibenzyl ester p-toluenesulfonate, 1,4-dibenzyl 2s-2-aminobutanedioate; para-toluene sulfonate
PubChem CID 2808531
IUPAC Name 1,4-dibenzyl 2-aminobutanedioate; 4-methylbenzene-1-sulfonic acid
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 159°C
Color White
Quantity 5 g
Formula Weight 485.55
Percent Purity ≥97.0% (HPLC,N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibenzyl L-Aspartate p-Toluenesulfonate