missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[3.2.2]nonane, 95%

Catalog No. AC438900050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438900050 5 g Glass bottle
AC438900010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438900050 Supplier Thermo Scientific Chemicals Supplier No. 438900050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 283-38-5 | C7H14N2 | 126.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 283-38-5
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.2
InChI Key XJKNACDCUAFDHD-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 3.2.2 nonane, 1,4-diazobicylco 3.2.2 nonane, 1,4-diaza-bicyclo 3.2.2 nonane, 1,4-diazabicyclo-3.2.2 nonane, 1,4-diazabicyclo 3.2.2 _nonane, 1,4-diazabicyclo 3.2.2-nonane
PubChem CID 398407
IUPAC Name 1,4-diazabicyclo[3.2.2]nonane
SMILES C1CN2CCC1NCC2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 58.0°C to 79.0°C
Color Colorless to Yellow
Boiling Point 77.0°C to 79.0°C (6.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 126.2
Percent Purity 95%
Physical Form Waxy Solid
Chemical Name or Material 1,4-Diazabicyclo[3.2.2]nonane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only