missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane 98.0+%, TCI America™

Catalog No. p-7126053
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
D013425G 25 g
D0134100G 100 g
D0134500G 500 g
3 options

Catalog No. D013425G

Supplier: TCI America D013425G

Only null left

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

Melting Point 155°C
Color White-Yellow
Boiling Point 174°C
Quantity 25 g
UN Number 3263
Formula Weight 112.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane
EINECSNumber (5)-1141
RTECSNumber HM0354200
TSCA Yes