missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1,4-Diazabicyclo[2.2.2]Octane

Catalog No. D278022KG
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D278022KG 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D278022KG Supplier Sigma Aldrich Supplier No. D278022KG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Diazabicyclo[2.2.2]Octane

Specifications

CAS 280-57-9
Melting Point 156°C to 159°C
Density 1.02 g/mL at 25°C
Molecular Formula C6H12N2
Linear Formula C6H12N2
Refractive Index n20/D 1.4634
MDL Number MFCD00006689
Quantity 25 g
Synonym TED; Triethylenediamine
Molecular Weight (g/mol) 112.17 g/mol