missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane 98.0+%, TCI America™

Catalog No. D023925G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D023925G 25 g
D0239400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D023925G Supplier TCI America Supplier No. D023925G

Chemical Identifiers

CAS 110-60-1
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008235
InChI Key KIDHWZJUCRJVML-UHFFFAOYSA-N
Synonym 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina
PubChem CID 1045
ChEBI CHEBI:17148
IUPAC Name butane-1,4-diamine
SMILES C(CCN)CN

Specifications

Color White
Boiling Point 159°C
Quantity 25 g
UN Number 2925
Formula Weight 88.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,4-Diaminobutane
EINECSNumber (2)-0152
RTECSNumber EJ6800000
TSCA Yes