missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane, 99%

Catalog No. AC112125000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112125000 500 g Glass bottle
AC112120250 25 g Glass bottle
AC112121000 100 g Glass bottle
AC112120025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC112125000 Supplier Thermo Scientific Chemicals Supplier No. 112125000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-60-1 | C4H12N2 | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-60-1
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008235
InChI Key KIDHWZJUCRJVML-UHFFFAOYSA-N
Synonym 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina
PubChem CID 1045
ChEBI CHEBI:17148
IUPAC Name butane-1,4-diamine
SMILES C(CCN)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 27.0°C
Color Colorless to Yellow
Density 0.8770g/mL
Boiling Point 158.0°C to 160.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.4610
Linear Formula H2N(CH2)4NH2
Quantity 500 g
Beilstein 04,264
Merck Index 15,8058
Specific Gravity 0.877
Solubility Information Solubility in water: 40g/L (20°C).
Formula Weight 88.15
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 1, 4-Diaminobutane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 203-782-3
RTECSNumber EJ6800000
TSCA TSCA

RUO – Research Use Only