Learn More
1,4-Diaminobutane, 98+%
CAS: 110-60-1 | C4H12N2 | 88.154 g/mol
Supplier: Thermo Scientific Chemicals B2131622
Description
1,4-Diaminobutane is used as a precursor in many biological systems and synthon for amido-ureas. It is involved in the synthesis of nylon 46 by reacting with adipic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Diaminobutane is used as a precursor in many biological systems and synthon for amido-ureas. It is involved in the synthesis of nylon 46 by reacting with adipic acid.
Solubility
Soluble in ether and alcohol.
Notes
Incompatible with acids, strong oxidizing agents, aldehydes, acid anhydrides and acid chlorides.
Specifications
| 1,4-Diaminobutane | |
| 25°C to 28°C | |
| 158°C to 160°C | |
| C4H12N2 | |
| H2N(CH2)4NH2 | |
| 100 g | |
| 605282 | |
| 14,7947 | |
| Soluble in ether and alcohol. | |
| C(CCN)CN | |
| 88.154 | |
| CHEBI:17148 | |
| ≥98% |
| 110-60-1 | |
| 0.877 | |
| 63°C (145°F) | |
| 1.4569 | |
| MFCD00008235 | |
| UN2928 | |
| Air sensitive; Hygroscopic | |
| 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina | |
| KIDHWZJUCRJVML-UHFFFAOYSA-N | |
| butane-1,4-diamine | |
| 1045 | |
| 88.15 |
Chemical Identifiers
| 110-60-1 | |
| 88.154 | |
| KIDHWZJUCRJVML-UHFFFAOYSA-N | |
| 1045 | |
| butane-1,4-diamine |
| C4H12N2 | |
| MFCD00008235 | |
| 1,4-diaminobutane, putrescine, 1,4-butanediamine, tetramethylenediamine, butylenediamine, putrescin, 1,4-butylenediamine, tetramethyldiamine, 1,4-tetramethylenediamine, putrescina | |
| CHEBI:17148 | |
| C(CCN)CN |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H311-H314-H330-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 203-782-3
RTECSNumber : EJ6800000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only