missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diaminoanthraquinone 97.0+%, TCI America™
$177.08 - $1002.86
Chemical Identifiers
| CAS | 128-95-0 |
|---|---|
| Molecular Formula | C14H10N2O2 |
| Molecular Weight (g/mol) | 238.25 |
| MDL Number | MFCD00001224 |
| InChI Key | FBMQNRKSAWNXBT-UHFFFAOYSA-N |
| Synonym | 1,4-diaminoanthraquinone, disperse violet 1, krisolamine, 9,10-anthracenedione, 1,4-diamino, grasol violet r, seacyl violet r, acetate red violet r, 1,4-diamino anthraquinone, setacyl violet r, oil violet r |
| PubChem CID | 31420 |
| IUPAC Name | 1,4-diamino-9,10-dihydroanthracene-9,10-dione |
| SMILES | NC1=CC=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O |
Chemical Identifiers
| 128-95-0 | |
| 238.25 | |
| FBMQNRKSAWNXBT-UHFFFAOYSA-N | |
| 31420 | |
| NC1=CC=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O |
| C14H10N2O2 | |
| MFCD00001224 | |
| 1,4-diaminoanthraquinone, disperse violet 1, krisolamine, 9,10-anthracenedione, 1,4-diamino, grasol violet r, seacyl violet r, acetate red violet r, 1,4-diamino anthraquinone, setacyl violet r, oil violet r | |
| 1,4-diamino-9,10-dihydroanthracene-9,10-dione |
Specifications
| 128-95-0 | |
| Black-Purple | |
| MFCD00001224 | |
| 1,4-diaminoanthraquinone, disperse violet 1, krisolamine, 9,10-anthracenedione, 1,4-diamino, grasol violet r, seacyl violet r, acetate red violet r, 1,4-diamino anthraquinone, setacyl violet r, oil violet r | |
| NC1=CC=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O | |
| 238.25 | |
| 238.25 | |
| Crystalline Powder |
| 270°C | |
| C14H10N2O2 | |
| 1 g | |
| FBMQNRKSAWNXBT-UHFFFAOYSA-N | |
| 1,4-diamino-9,10-dihydroanthracene-9,10-dione | |
| 31420 | |
| ≥97.0% (N) | |
| 1,4-Diaminoanthraquinone |
Safety and Handling
EINECSNumber : (4)-0710&(5)-2540
RTECSNumber : CB6300000
TSCA : Yes