missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diaminoanthraquinone 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 270°C |
| Color | Black-Purple |
| Quantity | 1 g |
| Formula Weight | 238.25 |
| Percent Purity | ≥97.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4-Diaminoanthraquinone |
Safety and Handling
| EINECSNumber | (4)-0710&(5)-2540 |
| RTECSNumber | CB6300000 |
| TSCA | Yes |