missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diamino-2,3-dichloroanthraquinone 93.0+%, TCI America™

Catalog No. D272525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D272525G 25 g
D27255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D272525G Supplier TCI America Supplier No. D272525G

Chemical Identifiers

CAS 81-42-5
Molecular Formula C14H8Cl2N2O2
Molecular Weight (g/mol) 307.13
MDL Number MFCD00035693
InChI Key KZYAYVSWIPZDKL-UHFFFAOYSA-N
Synonym Disperse Violet 28
PubChem CID 65731
IUPAC Name 1,4-diamino-2,3-dichloro-9,10-dihydroanthracene-9,10-dione
SMILES NC1=C(Cl)C(Cl)=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O

Specifications

Color Black-Purple
Quantity 25 g
Formula Weight 307.13
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diamino-2,3-dichloroanthraquinone
EINECSNumber (4)-0693&(5)-2549
TSCA Yes