missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetylbenzene 99.0+%, TCI America™

Catalog No. p-7126008
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00665G 5 g
D006625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00665G Supplier TCI America Supplier No. D00665G
Only null left

Chemical Identifiers

CAS 1009-61-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008750
InChI Key SKBBQSLSGRSQAJ-UHFFFAOYSA-N
Synonym 1,4-diacetylbenzene, 1,1'-1,4-phenylene diethanone, p-diacetylbenzene, p-acetylacetophenone, benzene, p-diacetyl, ethanone, 1,1'-1,4-phenylene bis, 1-4-acetylphenyl ethanone, 1-4-acetylphenyl ethan-1-one, 1,1-1,4-phenylene bis-ethanone, 4-acetylacetophenone
PubChem CID 13888
IUPAC Name 1-(4-acetylphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 113°C
Color White
Boiling Point 120°C
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diacetylbenzene