missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diacetoxybutane 98.0+%, TCI America™

Catalog No. p-7118938
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B123225G 25 g
B1232500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B123225G Supplier TCI America Supplier No. B123225G

Chemical Identifiers

CAS 628-67-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00014984
InChI Key XUKSWKGOQKREON-UHFFFAOYSA-N
Synonym 1,4-diacetoxybutane, 1,4-butanediol diacetate, tetramethylene diacetate, 1,4-butanediol, diacetate, butylene glycol diacetate, tetramethylene acetate, 1,4-butylene glycol diacetate, 1,4-butanediol, 1,4-diacetate, butane-1,4-diol diacetate, 1,4-butylene diacetate
PubChem CID 69410
ChEBI CHEBI:87345
IUPAC Name 4-acetyloxybutyl acetate
SMILES CC(=O)OCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 230°C
Quantity 25 g
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Diacetoxybutane
EINECSNumber (2)-2538
TSCA Yes