missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diacetoxybutane 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 230°C |
| Quantity | 500 g |
| Formula Weight | 174.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,4-Diacetoxybutane |
Safety and Handling
| EINECSNumber | (2)-2538 |
| TSCA | Yes |